missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(9H-Carbazol-9-yl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$134.21 - $134.21
Specifications
| CAS | 419536-33-7 |
|---|---|
| Melting Point | 264°C |
| Color | White |
| Molecular Formula | C18H14BNO2 |
| MDL Number | MFCD13176534 |
Specifications
| 419536-33-7 | |
| White | |
| MFCD13176534 | |
| 23120827 | |
| Crystalline Powder |
| 264°C | |
| C18H14BNO2 | |
| JGAVTCVHDMOQTJ-UHFFFAOYSA-N | |
| 287.13 |