accessibility menu, dialog, popup

UserName

CAS RN 419536-33-7

IUPAC Name: [4-(9H-carbazol-9-yl)phenyl]boronic acid
Synonyms: 4-(carbazol-9-yl)phenylboronic acid
Molecular Weight (g/mol): 287.13
Molecular Formula: C18H14BNO2
InChi Key: JGAVTCVHDMOQTJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Physical Form 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)