missing translation for 'onlineSavingsMsg'
Learn More

4-(9-Carbazolyl)benzeneboronic acid, 98%

CAS: 419536-33-7 | C18H14BNO2 | 287.13 g/mol

$148.64 - $543.71

Chemical Identifiers

CAS 419536-33-7
Molecular Formula C18H14BNO2
Molecular Weight (g/mol) 287.13
MDL Number MFCD13176534
InChI Key JGAVTCVHDMOQTJ-UHFFFAOYSA-N
Synonym 4-9h-carbazol-9-yl phenyl boronic acid, 4-9h-carbozol-9-yl phenylboronic acid, 4-9h-carbazol-9-yl phenylboronic acid, 4-9-carbazolyl benzeneboronic acid, 4-9h-9-carbozale phenylboronic acid, 4-9-carbazolyl phenylboronic acid, 4-carbazol-9-yl phenylboronic acid, boronic acid, 4-9h-carbazol-9-yl phenyl, amtb338
PubChem CID 23120827
SMILES OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6470303
View Documents
Thermo Scientific Chemicals
H6470303
1 g
Each for $148.64
Only null left
 
AAH6470306
View Documents
Thermo Scientific Chemicals
H6470306
5 g
Each for $543.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

419536-33-7
287.13
JGAVTCVHDMOQTJ-UHFFFAOYSA-N
23120827
C18H14BNO2
MFCD13176534
4-9h-carbazol-9-yl phenyl boronic acid, 4-9h-carbozol-9-yl phenylboronic acid, 4-9h-carbazol-9-yl phenylboronic acid, 4-9-carbazolyl benzeneboronic acid, 4-9h-9-carbozale phenylboronic acid, 4-9-carbazolyl phenylboronic acid, 4-carbazol-9-yl phenylboronic acid, boronic acid, 4-9h-carbazol-9-yl phenyl, amtb338
OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

419536-33-7
C18H14BNO2
1 g
4-9h-carbazol-9-yl phenyl boronic acid, 4-9h-carbozol-9-yl phenylboronic acid, 4-9h-carbazol-9-yl phenylboronic acid, 4-9-carbazolyl benzeneboronic acid, 4-9h-9-carbozale phenylboronic acid, 4-9-carbazolyl phenylboronic acid, 4-carbazol-9-yl phenylboronic acid, boronic acid, 4-9h-carbazol-9-yl phenyl, amtb338
OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
287.13
287.13
Powder
White
MFCD13176534
10565612
JGAVTCVHDMOQTJ-UHFFFAOYSA-N
(4-carbazol-9-ylphenyl)boronic acid
23120827
98%
4-(9-Carbazolyl)benzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only