missing translation for 'onlineSavingsMsg'
Learn More

4-(9-Carbazolyl)benzeneboronic acid, 98%

Catalog No. AAH6470303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6470303 1 g
AAH6470306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6470303 Supplier Thermo Scientific Chemicals Supplier No. H6470303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 419536-33-7 | C18H14BNO2 | 287.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 419536-33-7
Molecular Formula C18H14BNO2
Molecular Weight (g/mol) 287.13
MDL Number MFCD13176534
InChI Key JGAVTCVHDMOQTJ-UHFFFAOYSA-N
Synonym 4-9h-carbazol-9-yl phenyl boronic acid, 4-9h-carbozol-9-yl phenylboronic acid, 4-9h-carbazol-9-yl phenylboronic acid, 4-9-carbazolyl benzeneboronic acid, 4-9h-9-carbozale phenylboronic acid, 4-9-carbazolyl phenylboronic acid, 4-carbazol-9-yl phenylboronic acid, boronic acid, 4-9h-carbazol-9-yl phenyl, amtb338
PubChem CID 23120827
SMILES OB(O)C1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Color White
Quantity 1 g
Beilstein 10565612
IUPAC Name (4-carbazol-9-ylphenyl)boronic acid
Formula Weight 287.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-(9-Carbazolyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only