missing translation for 'onlineSavingsMsg'
Learn More

4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one 98.0+%, TCI America™

Catalog No. p-7137233
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24711G 1 g
T24715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24711G Supplier TCI America Supplier No. T24711G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 632339-23-2
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD10566937
InChI Key UAFDSOCDQRHNCL-UHFFFAOYNA-N
Synonym 2,6-Epoxy-2,2a,5,5a,6,6a-hexahydrooxireno[f]isobenzofuran-3(1aH)-one
PubChem CID 20632532
IUPAC Name 4,9,11-trioxatetracyclo[5.3.1.0²,⁶.0⁸,¹⁰]undecan-3-one
SMILES O=C1OCC2C3OC(C4OC34)C12

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 1 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one