accessibility menu, dialog, popup

UserName

CAS RN 632339-23-2

IUPAC Name: 4,9,11-trioxatetracyclo[5.3.1.0²,⁶.0⁸,¹⁰]undecan-3-one
Synonyms: 4,9,11-trioxatetracyclo[5.3.1.0²,⁶.0⁸,¹⁰]undecan-3-one
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: UAFDSOCDQRHNCL-UHFFFAOYNA-N
SMILES: O=C1OCC2C3OC(C4OC34)C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results