missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,8-Bis-n-octyloxybenzo[1,2-b:4,5-b']dithiophene 98.0+%, TCI America™
$1039.59 - $1039.59
Chemical Identifiers
| CAS | 1098102-94-3 |
|---|---|
| Molecular Formula | C26H38O2S2 |
| Molecular Weight (g/mol) | 446.71 |
| MDL Number | MFCD26793498 |
| InChI Key | LWSGUMOETOXOHL-UHFFFAOYSA-N |
| PubChem CID | 58280899 |
| IUPAC Name | 2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,â·]dodeca-1,3(7),5,8,11-pentaene |
| SMILES | CCCCCCCCOC1=C2C=CSC2=C(OCCCCCCCC)C2=C1SC=C2 |
Chemical Identifiers
| 1098102-94-3 | |
| 446.71 | |
| LWSGUMOETOXOHL-UHFFFAOYSA-N | |
| 2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,â·]dodeca-1,3(7),5,8,11-pentaene |
| C26H38O2S2 | |
| MFCD26793498 | |
| 58280899 | |
| CCCCCCCCOC1=C2C=CSC2=C(OCCCCCCCC)C2=C1SC=C2 |
Specifications
| 1098102-94-3 | |
| White-Yellow | |
| MFCD26793498 | |
| LWSGUMOETOXOHL-UHFFFAOYSA-N | |
| 2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,â·]dodeca-1,3(7),5,8,11-pentaene | |
| 58280899 | |
| ≥98.0% (GC) | |
| 4,8-Bis-n-octyloxybenzo[1,2-b:4,5-b′]dithiophene |
| 48°C | |
| C26H38O2S2 | |
| 1 g | |
| CCCCCCCCOC1=C2C=CSC2=C(OCCCCCCCC)C2=C1SC=C2 | |
| 446.71 | |
| 446.71 | |
| Crystalline Powder |
Safety and Handling
TSCA : No