missing translation for 'onlineSavingsMsg'
Learn More

4,8-Bis-n-octyloxybenzo[1,2-b:4,5-b']dithiophene 98.0+%, TCI America™

Catalog No. b43961g
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
B43961G 1 g
1 options

Catalog No. B43961G

Supplier: TCI America B43961G

Only null left

Chemical Identifiers

CAS 1098102-94-3
Molecular Formula C26H38O2S2
Molecular Weight (g/mol) 446.71
MDL Number MFCD26793498
InChI Key LWSGUMOETOXOHL-UHFFFAOYSA-N
PubChem CID 58280899
IUPAC Name 2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
SMILES CCCCCCCCOC1=C2C=CSC2=C(OCCCCCCCC)C2=C1SC=C2

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 1 g
Formula Weight 446.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,8-Bis-n-octyloxybenzo[1,2-b:4,5-b′]dithiophene