missing translation for 'onlineSavingsMsg'
Learn More

4,8-Bis[5-(2-ethylhexyl)thiophen-2-yl]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene 96.0+%, TCI America™

Catalog No. B4437200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4437200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4437200MG Supplier TCI America Supplier No. B4437200MG
Only null left

Chemical Identifiers

CAS 1352642-37-5
Molecular Formula C40H58S4Sn2
Molecular Weight (g/mol) 904.564
InChI Key OCFFMJYHZKHRKM-UHFFFAOYSA-N
PubChem CID 90187248
IUPAC Name [4,8-bis[5-(2-ethylhexyl)thiophen-2-yl]-2-trimethylstannylthieno[2,3-f][1]benzothiol-6-yl]-trimethylstannane
SMILES CCCCC(CC)CC1=CC=C(S1)C2=C3C=C(SC3=C(C4=C2SC(=C4)[Sn](C)(C)C)C5=CC=C(S5)CC(CC)CCCC)[Sn](C)(C)C

Specifications

Color Yellow
Quantity 200 mg
UN Number 3146
Formula Weight 904.56
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,8-Bis[5-(2-ethylhexyl)thiophen-2-yl]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b′]dithiophene