accessibility menu, dialog, popup

UserName

CAS RN 1352642-37-5

CH3CH3SSSSnH3CCH3CH3SCH3CH3SnCH3CH3CH3

CAS RN 1352642-37-5

IUPAC Name: {2,8-bis[5-(2-ethylhexyl)thiophen-2-yl]-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl}trimethylstannane
Synonyms: {2,8-bis[5-(2-ethylhexyl)thiophen-2-yl]-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl}trimethylstannane
Molecular Weight (g/mol): 904.56
Molecular Formula: C40H58S4Sn2
InChi Key: OCFFMJYHZKHRKM-UHFFFAOYNA-N
SMILES: CCCCC(CC)CC1=CC=C(S1)C1=C2C=C(SC2=C(C2=CC=C(CC(CC)CCCC)S2)C2=C1SC(=C2)[Sn](C)(C)C)[Sn](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results