missing translation for 'onlineSavingsMsg'
Learn More

4,8-Bis(2-butyl-n-octyloxy)-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene 97.0+%, TCI America™

Catalog No. B4685200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4685200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4685200MG Supplier TCI America Supplier No. B4685200MG
Only null left

Chemical Identifiers

CAS 1271439-08-7
Molecular Formula C40H70O2S2Sn2
Molecular Weight (g/mol) 884.54
MDL Number MFCD28144735
InChI Key SWDLPBRTONKXQI-UHFFFAOYNA-N
PubChem CID 67514894
IUPAC Name {2,8-bis[(2-butyloctyl)oxy]-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl}trimethylstannane
SMILES CCCCCCC(CCCC)COC1=C2C=C(SC2=C(OCC(CCCC)CCCCCC)C2=C1SC(=C2)[Sn](C)(C)C)[Sn](C)(C)C

Specifications

Quantity 200 mg
UN Number 3146
Formula Weight 884.54
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,8-Bis(2-butyl-n-octyloxy)-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b′]dithiophene