accessibility menu, dialog, popup

UserName

CAS RN 1271439-08-7

H3CH3COSSSnCH3CH3H3COCH3CH3SnH3CCH3H3C

CAS RN 1271439-08-7

IUPAC Name: {2,8-bis[(2-butyloctyl)oxy]-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl}trimethylstannane
Synonyms: {2,8-bis[(2-butyloctyl)oxy]-11-(trimethylstannyl)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),2,5,7,11-pentaen-5-yl}trimethylstannane
Molecular Weight (g/mol): 884.54
Molecular Formula: C40H70O2S2Sn2
InChi Key: SWDLPBRTONKXQI-UHFFFAOYNA-N
SMILES: CCCCCCC(CCCC)COC1=C2C=C(SC2=C(OCC(CCCC)CCCCCC)C2=C1SC(=C2)[Sn](C)(C)C)[Sn](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results