missing translation for 'onlineSavingsMsg'
Learn More

4',7-Dimethoxyisoflavone 97.0+%, TCI America™

Catalog No. d43895g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43895G 5 g
D43891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43895G Supplier TCI America Supplier No. D43895G

Chemical Identifiers

CAS 1157-39-7
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00075889
InChI Key LPNBCGIVZXHHHO-UHFFFAOYSA-N
Synonym 4',7-dimethoxyisoflavone, 7,4'-dimethoxyisoflavone, 7-methoxy-3-4-methoxyphenyl-4h-chromen-4-one, 7,4'-di-o-methyldaidzein, unii-f5547dw3co, 7-methoxy-3-4-methoxyphenyl chromen-4-one, daidzein dimethyl ether, spectrum_000697, specplus_000113, spectrum2_000185
PubChem CID 136419
IUPAC Name 7-methoxy-3-(4-methoxyphenyl)chromen-4-one
SMILES COC1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)OC

Specifications

Melting Point 163°C
Color White-Yellow
Quantity 5 g
Formula Weight 282.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′,7-Dimethoxyisoflavone