missing translation for 'onlineSavingsMsg'
Learn More

4',7-Dimethoxyisoflavone, 97%

Catalog No. p-7046294
Change view
Click to view available options
Quantity:
100 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13838MC 100 mg
AAL1383804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13838MC Supplier Thermo Scientific Chemicals Supplier No. L13838MC
Only null left

CAS: 1157-39-7 | C17H14O4 | 282.295 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1157-39-7
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00075889
InChI Key LPNBCGIVZXHHHO-UHFFFAOYSA-N
Synonym 4',7-dimethoxyisoflavone, 7,4'-dimethoxyisoflavone, 7-methoxy-3-4-methoxyphenyl-4h-chromen-4-one, 7,4'-di-o-methyldaidzein, unii-f5547dw3co, 7-methoxy-3-4-methoxyphenyl chromen-4-one, daidzein dimethyl ether, spectrum_000697, specplus_000113, spectrum2_000185
PubChem CID 136419
IUPAC Name 7-methoxy-3-(4-methoxyphenyl)chromen-4-one
SMILES COC1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)OC

Specifications

Melting Point 161°C to 163°C
Quantity 100 mg
Beilstein 265550
Formula Weight 282.3
Percent Purity 97%
Chemical Name or Material 4',7-Dimethoxyisoflavone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only