missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dichloroquinoline 98.0+%, TCI America™

Catalog No. D110025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D110025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D110025G Supplier TCI America Supplier No. D110025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 86-98-6
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.046
MDL Number MFCD00006774
InChI Key HXEWMTXDBOQQKO-UHFFFAOYSA-N
Synonym quinoline, 4,7-dichloro, 4,7-dichloro-quinoline, unii-z61o2heq2j, z61o2heq2j, 4,7dichloroquinoline, pubchem5880, 4.7-dichloroquinoline, 4,7 dichloroquinoline, quinoline,7-dichloro, amodiaquine impurity 1
PubChem CID 6866
IUPAC Name 4,7-dichloroquinoline
SMILES C1=CC2=C(C=CN=C2C=C1Cl)Cl

Specifications

Melting Point 87°C
Color White-Yellow
Boiling Point 148°C
Quantity 25 g
Formula Weight 198.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dichloroquinoline
RTECSNumber VB4200000
TSCA Yes