missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dichloroquinoline, 98%

Catalog No. AC113751000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113751000 100 g Glass bottle
AC113750250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113751000 Supplier Thermo Scientific Chemicals Supplier No. 113751000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-98-6 | C9H5Cl2N | 198.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86-98-6
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.05
MDL Number MFCD00006774
InChI Key HXEWMTXDBOQQKO-UHFFFAOYSA-N
PubChem CID 6866
IUPAC Name 4,7-dichloroquinoline
SMILES C1=CC2=C(C=CN=C2C=C1Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 84°C to 86°C
Color Brown to White
Boiling Point 148°C
Flash Point 164°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Synonym quinoline, 4,7-dichloro, 4,7-dichloro-quinoline, unii-z61o2heq2j, z61o2heq2j, 4,7dichloroquinoline, pubchem5880, 4.7-dichloroquinoline, 4,7 dichloroquinoline, quinoline,7-dichloro, amodiaquine impurity 1
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid,174g/L toluene ( 20 c),soluble in acetone,methanol and ethanol
Formula Weight 198.05
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4, 7-Dichloroquinoline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-714-7
RTECSNumber VB4200000
TSCA TSCA

RUO – Research Use Only