missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dichloroquinoline, 98%

Catalog No. AAA1598514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1598514 25 g
AAA1598522 100 g
AAA1598536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1598514 Supplier Thermo Scientific Chemicals Supplier No. A1598514

CAS: 86-98-6 | C9H5Cl2N | 198.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4,7-Dichloroquinoline is used as a drug chloroquine phosphate intermediate. It is also is used in the synthesis of hybrid aminoquinoline-triazine derivatives that show anti-microbial activity. In addition, it is used in the synthesis of novel oxazolidinones as anti-microbial agents showing efficacy against common bacterial strains.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 86-98-6
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.046
MDL Number MFCD00006774
InChI Key HXEWMTXDBOQQKO-UHFFFAOYSA-N
PubChem CID 6866
IUPAC Name 4,7-dichloroquinoline
SMILES C1=CC2=C(C=CN=C2C=C1Cl)Cl

Specifications

Melting Point 83°C to 87°C
Boiling Point 148°C (10 mmHg)
Flash Point 164°C (327°F)
Quantity 25 g
Beilstein 125359
Synonym quinoline, 4,7-dichloro, 4,7-dichloro-quinoline, unii-z61o2heq2j, z61o2heq2j, 4,7dichloroquinoline, pubchem5880, 4.7-dichloroquinoline, 4,7 dichloroquinoline, quinoline,7-dichloro, amodiaquine impurity 1
Solubility Information Insoluble in water.
Formula Weight 198.05
Percent Purity 98%
Chemical Name or Material 4,7-Dichloroquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-714-7
RTECSNumber VB4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only