missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole 98.0+%, TCI America™
Supplier: TCI America D47501G
Specifications
| 4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole | |
| 162°C | |
| C6HBr2FN2S | |
| 1 g | |
| FC1=C(Br)C2=NSN=C2C(Br)=C1 | |
| 311.95 | |
| 311.95 | |
| Crystalline Powder |
| 1347736-74-6 | |
| White-Yellow | |
| MFCD26939241 | |
| KVZDYOVYIHJETJ-UHFFFAOYSA-N | |
| 4,7-dibromo-5-fluoro-2,1,3-benzothiadiazole | |
| 67515326 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1347736-74-6 | |
| 311.95 | |
| KVZDYOVYIHJETJ-UHFFFAOYSA-N | |
| 4,7-dibromo-5-fluoro-2,1,3-benzothiadiazole |
| C6HBr2FN2S | |
| MFCD26939241 | |
| 67515326 | |
| FC1=C(Br)C2=NSN=C2C(Br)=C1 |
Safety and Handling
TSCA : No