missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole 98.0+%, TCI America™

Catalog No. p-7128439
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4750200MG 200 mg
D47501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4750200MG Supplier TCI America Supplier No. D4750200MG
Only null left

Chemical Identifiers

CAS 1347736-74-6
Molecular Formula C6HBr2FN2S
Molecular Weight (g/mol) 311.95
MDL Number MFCD26939241
InChI Key KVZDYOVYIHJETJ-UHFFFAOYSA-N
PubChem CID 67515326
IUPAC Name 4,7-dibromo-5-fluoro-2,1,3-benzothiadiazole
SMILES FC1=C(Br)C2=NSN=C2C(Br)=C1

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 200 mg
Formula Weight 311.95
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-5-fluoro-2,1,3-benzothiadiazole