missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,7-Dibromo-2,1,3-benzothiadiazole 98.0+%, TCI America™
Supplier: TCI America D38425G
Specifications
| 4,7-Dibromo-2,1,3-benzothiadiazole | |
| 189°C | |
| C6H2Br2N2S | |
| 5 g | |
| FEOWHLLJXAECMU-UHFFFAOYSA-N | |
| 4,7-dibromo-2,1,3-benzothiadiazole | |
| 626361 | |
| ≥98.0% (GC) |
| 15155-41-6 | |
| White-Yellow | |
| MFCD00658844 | |
| 4,7-dibromobenzo c 1,2,5 thiadiazole, 4,7-dibromo-2,1,3-benzothidiazole, 4,7-dibromobenzo c-1,2,5-thiadiazole, 2,1,3-benzothiadiazole, 4,7-dibromo, acmc-1bu18, ksc490o9b, 4,7-dibromobenzo 1,2,5 thiadiazole, 4,7-dibromobenzo 2,1,3 thiadiazole, 4,7-dibromo-2,1,3-benzothiadiazole, 4,7-dibromo-benzo 1,2,5 thiadiazole | |
| BrC1=CC=C(Br)C2=NSN=C12 | |
| 293.96 | |
| 293.96 | |
| Crystalline Powder |
Chemical Identifiers
| 15155-41-6 | |
| 293.96 | |
| FEOWHLLJXAECMU-UHFFFAOYSA-N | |
| 626361 | |
| BrC1=CC=C(Br)C2=NSN=C12 |
| C6H2Br2N2S | |
| MFCD00658844 | |
| 4,7-dibromobenzo c 1,2,5 thiadiazole, 4,7-dibromo-2,1,3-benzothidiazole, 4,7-dibromobenzo c-1,2,5-thiadiazole, 2,1,3-benzothiadiazole, 4,7-dibromo, acmc-1bu18, ksc490o9b, 4,7-dibromobenzo 1,2,5 thiadiazole, 4,7-dibromobenzo 2,1,3 thiadiazole, 4,7-dibromo-2,1,3-benzothiadiazole, 4,7-dibromo-benzo 1,2,5 thiadiazole | |
| 4,7-dibromo-2,1,3-benzothiadiazole |
Safety and Handling
TSCA : No