accessibility menu, dialog, popup

UserName

CAS RN 15155-41-6

IUPAC Name: 4,7-dibromo-2,1,3-benzothiadiazole
Synonyms: 4,7-dibromo-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 293.96
Molecular Formula: C6H2Br2N2S
InChi Key: FEOWHLLJXAECMU-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)C2=NSN=C12
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)