missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dibromo-2,1,3-benzothiadiazole 98.0+%, TCI America™

Catalog No. D38421G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38421G 1 g
D38425G 5 g
D384225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D38421G Supplier TCI America Supplier No. D38421G

Chemical Identifiers

CAS 15155-41-6
Molecular Formula C6H2Br2N2S
Molecular Weight (g/mol) 293.96
MDL Number MFCD00658844
InChI Key FEOWHLLJXAECMU-UHFFFAOYSA-N
Synonym 4,7-dibromobenzo c 1,2,5 thiadiazole, 4,7-dibromo-2,1,3-benzothidiazole, 4,7-dibromobenzo c-1,2,5-thiadiazole, 2,1,3-benzothiadiazole, 4,7-dibromo, acmc-1bu18, ksc490o9b, 4,7-dibromobenzo 1,2,5 thiadiazole, 4,7-dibromobenzo 2,1,3 thiadiazole, 4,7-dibromo-2,1,3-benzothiadiazole, 4,7-dibromo-benzo 1,2,5 thiadiazole
PubChem CID 626361
IUPAC Name 4,7-dibromo-2,1,3-benzothiadiazole
SMILES BrC1=CC=C(Br)C2=NSN=C12

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 1 g
Formula Weight 293.96
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-2,1,3-benzothiadiazole