missing translation for 'onlineSavingsMsg'
Learn More

4,7-Dibromo-2,1,3-benzothiadiazole, 97%

Catalog No. p-7037546
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5630603 1 g
AAH5630606 5 g
AAH5630614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5630603 Supplier Thermo Scientific Chemicals Supplier No. H5630603
Only null left

CAS: 15155-41-6 | C6H2Br2N2S | 293.96 g/mol

4,7-Dibromo-2,1,3-benzothiadiazole is the building block or monomer for the synthesis of light-emitting diodes and conducting polymers for organic electronics. It is used as an intermediate for the synthesis of poly[N-9'-heptadecanyl-2,7-carbazole-alt-5,5-(4',7'-di-2-thienyl-2',1',3'-benzothiadiazole)] (PCDTBT) and poly[2,1,3-benzothiadiazole-4,7-diyl[4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene-2,6-diyl]] (PCPDTBT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,7-Dibromo-2,1,3-benzothiadiazole is the building block or monomer for the synthesis of light-emitting diodes and conducting polymers for organic electronics. It is used as an intermediate for the synthesis of poly[N-9′-heptadecanyl-2,7-carbazole-alt-5,5-(4′,7′-di-2-thienyl-2′,1′,3′-benzothiadiazole)] (PCDTBT) and poly[2,1,3-benzothiadiazole-4,7-diyl[4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b′]dithiophene-2,6-diyl]] (PCPDTBT).

Solubility
Soluble in toluene.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 15155-41-6
Molecular Formula C6H2Br2N2S
Molecular Weight (g/mol) 293.96
MDL Number MFCD00658844
InChI Key FEOWHLLJXAECMU-UHFFFAOYSA-N
Synonym 4,7-dibromobenzo c 1,2,5 thiadiazole, 4,7-dibromo-2,1,3-benzothidiazole, 4,7-dibromobenzo c-1,2,5-thiadiazole, 2,1,3-benzothiadiazole, 4,7-dibromo, acmc-1bu18, ksc490o9b, 4,7-dibromobenzo 1,2,5 thiadiazole, 4,7-dibromobenzo 2,1,3 thiadiazole, 4,7-dibromo-2,1,3-benzothiadiazole, 4,7-dibromo-benzo 1,2,5 thiadiazole
PubChem CID 626361
IUPAC Name 4,7-dibromo-2,1,3-benzothiadiazole
SMILES BrC1=CC=C(Br)C2=NSN=C12

Specifications

Melting Point 186°C to 190°C
Quantity 1 g
UN Number UN2811
Solubility Information Soluble in toluene.
Formula Weight 293.97
Percent Purity 97%
Chemical Name or Material 4,7-Dibromo-2,1,3-benzothiadiazole
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only