missing translation for 'onlineSavingsMsg'
Learn More

4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate, 98%

Catalog No. AAJ6432606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6432606 5 g
AAJ6432614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6432606 Supplier Thermo Scientific Chemicals Supplier No. J6432606

CAS: 6398-98-7 | C20H28Cl3N3O3 | 464.812 g/mol

An anti-malarial quinoline derivative that acts as an histamine N-methyltransferase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

An anti-malarial quinoline derivative that acts as an histamine N-methyltransferase inhibitor.

Chemical Identifiers

CAS 6398-98-7
Molecular Formula C20H28Cl3N3O3
Molecular Weight (g/mol) 464.812
MDL Number MFCD00078857
InChI Key YVNAYSHNIILOJS-UHFFFAOYSA-N
Synonym amodiaquin dihydrochloride dihydrate, amodiaquine hcl, amodiaquine hydrochloride, amodiaquine dihydrochloride dihydrate, unii-k6pw2s574l, 4-7-chloroquinolin-4-yl amino-2-diethylamino methyl phenol dihydrochloride dihydrate, amodiaquine hydrochloride usp, cam-aq 1
PubChem CID 64646
ChEBI CHEBI:50652
IUPAC Name 4-[(7-chloroquinolin-4-yl)amino]-2-(diethylaminomethyl)phenol;dihydrate;dihydrochloride
SMILES CCN(CC)CC1=C(C=CC(=C1)NC2=C3C=CC(=CC3=NC=C2)Cl)O.O.O.Cl.Cl

Specifications

Color Yellow
Quantity 5 g
Merck Index 14,572
Solubility Information Soluble in DMSO or methanol
Formula Weight 464.82 (392.33 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-(7-Chloro-4-quinolinylamino)-2-(diethylaminomethyl)phenol dihydrochloride dihydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-706-0
RTECSNumber GO7300100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only