missing translation for 'onlineSavingsMsg'
Learn More

4,7-Bis(5-trimethylstannyl-2-thienyl)-2,1,3-benzothiadiazole 95.0+%, TCI America™

Catalog No. B4453200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4453200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4453200MG Supplier TCI America Supplier No. B4453200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1025451-57-3
Molecular Formula C20H24N2S3Sn2
Molecular Weight (g/mol) 626.026
InChI Key INHYKPMJJAWZKY-UHFFFAOYSA-N
PubChem CID 58153009
IUPAC Name trimethyl-[5-[4-(5-trimethylstannylthiophen-2-yl)-2,1,3-benzothiadiazol-7-yl]thiophen-2-yl]stannane
SMILES C[Sn](C)(C)C1=CC=C(S1)C2=CC=C(C3=NSN=C23)C4=CC=C(S4)[Sn](C)(C)C

Specifications

Quantity 200 mg
UN Number 3146
Formula Weight 626.03
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Bis(5-trimethylstannyl-2-thienyl)-2,1,3-benzothiadiazole