accessibility menu, dialog, popup

UserName

H3CSnH3CCH3SNSNSSnH3CH3CCH3

CAS RN 1025451-57-3

IUPAC Name: 4,7-bis[5-(trimethylstannyl)thiophen-2-yl]-2,1,3-benzothiadiazole
Synonyms: 4,7-bis[5-(trimethylstannyl)thiophen-2-yl]-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 626.03
Molecular Formula: C20H24N2S3Sn2
InChi Key: INHYKPMJJAWZKY-UHFFFAOYSA-N
SMILES: C[Sn](C)(C)C1=CC=C(S1)C1=CC=C(C2=CC=C(S2)[Sn](C)(C)C)C2=NSN=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results