missing translation for 'onlineSavingsMsg'
Learn More

4,7-Bis(5-bromo-2-thienyl)-2-n-octyl-2H-benzotriazole 90.0+%, TCI America™

Catalog No. B4542100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4542100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4542100MG Supplier TCI America Supplier No. B4542100MG
Only null left

Chemical Identifiers

CAS 1254062-41-3
Molecular Formula C22H23Br2N3S2
Molecular Weight (g/mol) 553.38
MDL Number MFCD27923045
InChI Key KPXJKVVGOPZOPK-UHFFFAOYSA-N
PubChem CID 71721485
IUPAC Name 4,7-bis(5-bromothiophen-2-yl)-2-octyl-2H-1,2,3-benzotriazole
SMILES CCCCCCCCN1N=C2C(=N1)C(=CC=C2C1=CC=C(Br)S1)C1=CC=C(Br)S1

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 553.38
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Bis(5-bromo-2-thienyl)-2-n-octyl-2H-benzotriazole