accessibility menu, dialog, popup

UserName

H3CNNNSBrSBr

CAS RN 1254062-41-3

IUPAC Name: 4,7-bis(5-bromothiophen-2-yl)-2-octyl-2H-1,2,3-benzotriazole
Synonyms: 4,7-bis(5-bromothiophen-2-yl)-2-octyl-1,2,3-benzotriazole
Molecular Weight (g/mol): 553.38
Molecular Formula: C22H23Br2N3S2
InChi Key: KPXJKVVGOPZOPK-UHFFFAOYSA-N
SMILES: CCCCCCCCN1N=C2C(=N1)C(=CC=C2C1=CC=C(Br)S1)C1=CC=C(Br)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results