missing translation for 'onlineSavingsMsg'
Learn More

4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole 95.0+%, TCI America™

Catalog No. p-7123022
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35731G 1 g
B35735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35731G Supplier TCI America Supplier No. B35731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 934365-16-9
Molecular Formula C18H26B2N2O4S
Molecular Weight (g/mol) 388.10
MDL Number MFCD11656086
InChI Key DHWADSHFLSDMBJ-UHFFFAOYSA-N
Synonym 2,1,3-Benzothiadiazole-4,7-diboronic Acid Bis(pinacol) Ester
PubChem CID 46739758
IUPAC Name 4,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(B2OC(C)(C)C(C)(C)O2)C2=NSN=C12

Specifications

Melting Point 214°C
Quantity 1 g
Formula Weight 388.10
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole