accessibility menu, dialog, popup

UserName

CAS RN 934365-16-9

IUPAC Name: 4,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole
Synonyms: 4,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 388.1
Molecular Formula: C18H26B2N2O4S
InChi Key: DHWADSHFLSDMBJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(B2OC(C)(C)C(C)(C)O2)C2=NSN=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results