missing translation for 'onlineSavingsMsg'
Learn More

4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane 98.0+%, TCI America™

Catalog No. H09325G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09325G 5 g
H09321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09325G Supplier TCI America Supplier No. H09325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23978-09-8
Molecular Formula C18H36N2O6
Molecular Weight (g/mol) 376.49
MDL Number MFCD00005111
InChI Key AUFVJZSDSXXFOI-UHFFFAOYSA-N
Synonym kryptofix 222, 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo 8.8.8 hexacosane, cryptating agent 222, cryptand 222, cryptofix 222, kriptofix 222, cryptate 222, ligand 222, kryptand 222, cryptand c 222
PubChem CID 72801
IUPAC Name 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
SMILES C1COCCN2CCOCCOCCN(CCO1)CCOCCOCC2

Specifications

Melting Point 71°C
Color White
Quantity 5 g
Formula Weight 376.49
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
RTECSNumber MP4750000
TSCA No