accessibility menu, dialog, popup

UserName

ONOCH2ONH2COOO

CAS RN 23978-09-8

IUPAC Name: 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
Synonyms: 2.2.2-cryptand cryptating agent 222
Molecular Weight (g/mol): 376.49
Molecular Formula: C18H36N2O6
InChi Key: AUFVJZSDSXXFOI-UHFFFAOYSA-N
SMILES: C1COCCN2CCOCCOCCN(CCO1)CCOCCOCC2
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)