missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4,6-O-Ethylidene-D-glucopyranose, 90%

Catalog No. p-7043252
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6658303 1 g
AAJ6658306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6658303 Supplier Thermo Scientific Chemicals Supplier No. J6658303
Only null left

CAS: 13224-99-2 | C8H14O6 | 206.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13224-99-2
Molecular Formula C8H14O6
Molecular Weight (g/mol) 206.19
MDL Number MFCD00006820
InChI Key VZPBLPQAMPVTFO-UHFFFAOYNA-N
Synonym 4,6-o-ethylidene-a-d-glucose
PubChem CID 21581900
IUPAC Name (2R,4aR,6S,7R,8R,8aS)-2-methyl-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxine-6,7,8-triol
SMILES CC1OCC2OC(O)C(O)C(O)C2O1

Specifications

Color White
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 206.19
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 4,6-O-Ethylidene-D-glucopyranose
EINECSNumber 236-198-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only