accessibility menu, dialog, popup

UserName

CAS RN 13224-99-2

IUPAC Name: 2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxine-6,7,8-triol
Synonyms: 2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxine-6,7,8-triol
Molecular Weight (g/mol): 206.19
Molecular Formula: C8H14O6
InChi Key: VZPBLPQAMPVTFO-UHFFFAOYNA-N
SMILES: CC1OCC2OC(O)C(O)C(O)C2O1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)