missing translation for 'onlineSavingsMsg'
Learn More

4,6-O-Benzylidene-D-glucal, 97%, Thermo Scientific Chemicals

$224.19 - $224.19

Chemical Identifiers

CAS 14125-70-3
Molecular Formula C13H14O4
Molecular Weight (g/mol) 234.25
MDL Number MFCD00167506
InChI Key XMDUTBYCCVWPLD-FVCCEPFGSA-N
Synonym 4,6-o-benzylidene-d-glucal, 2s,4ar,8r,8as-2-phenyl-4,4a,8,8a-tetrahydropyrano 3,2-d 1,3 dioxin-8-ol
PubChem CID 7067543
IUPAC Name (2S,4aR,8R,8aS)-2-phenyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
SMILES O[C@@H]1C=CO[C@@H]2CO[C@H](O[C@@H]12)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6025903
View Documents
Thermo Scientific Chemicals
H6025903
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14125-70-3
234.25
XMDUTBYCCVWPLD-FVCCEPFGSA-N
7067543
O[C@@H]1C=CO[C@@H]2CO[C@H](O[C@@H]12)C1=CC=CC=C1
C13H14O4
MFCD00167506
4,6-o-benzylidene-d-glucal, 2s,4ar,8r,8as-2-phenyl-4,4a,8,8a-tetrahydropyrano 3,2-d 1,3 dioxin-8-ol
(2S,4aR,8R,8aS)-2-phenyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol

Specifications

14125-70-3
MFCD00167506
4,6-o-benzylidene-d-glucal, 2s,4ar,8r,8as-2-phenyl-4,4a,8,8a-tetrahydropyrano 3,2-d 1,3 dioxin-8-ol
O[C@@H]1C=CO[C@@H]2CO[C@H](O[C@@H]12)C1=CC=CC=C1
234.25
234.25
4,6-O-Benzylidene-D-glucal
C13H14O4
1 g
XMDUTBYCCVWPLD-FVCCEPFGSA-N
(2S,4aR,8R,8aS)-2-phenyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
7067543
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only