missing translation for 'onlineSavingsMsg'
Learn More

4,6-O-Benzylidene-D-glucal, 97%, Thermo Scientific Chemicals

Catalog No. aah6025903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6025903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6025903 Supplier Thermo Scientific Chemicals Supplier No. H6025903
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14125-70-3
Molecular Formula C13H14O4
Molecular Weight (g/mol) 234.25
MDL Number MFCD00167506
InChI Key XMDUTBYCCVWPLD-FVCCEPFGSA-N
Synonym 4,6-o-benzylidene-d-glucal, 2s,4ar,8r,8as-2-phenyl-4,4a,8,8a-tetrahydropyrano 3,2-d 1,3 dioxin-8-ol
PubChem CID 7067543
IUPAC Name (2S,4aR,8R,8aS)-2-phenyl-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol
SMILES O[C@@H]1C=CO[C@@H]2CO[C@H](O[C@@H]12)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 234.25
Percent Purity 97%
Chemical Name or Material 4,6-O-Benzylidene-D-glucal
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only