missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-O-(4-Methoxybenzylidene)-D-glucal, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6063303
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4,6-O-(4-Methoxybenzylidene)-D-glucal is used as pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications
| 4,6-O-(4-Methoxybenzylidene)-D-glucal | |
| C14H16O5 | |
| 1 g | |
| SSKZQXMNNREVNP-HABKJSAYSA-N | |
| (4aR,8R,8aS)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol | |
| 46185760 | |
| 97% |
| 312623-79-3 | |
| MFCD01863528 | |
| 4,6-o-4-methoxybenzylidene-d-glucal, 4 6-o-4-methoxybenzylidene-d-glucal, 4-o,6-o-4-methoxybenzylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,5-anhydro-4-o,6-o-4-methoxybenzylidene-2-deoxy-d-arabino-hexa-1-enitol, 4ar,8r,8as-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol | |
| COC1=CC=C(C=C1)C2OCC3C(O2)C(C=CO3)O | |
| 264.277 | |
| 264.27 |
Chemical Identifiers
| 312623-79-3 | |
| 264.277 | |
| SSKZQXMNNREVNP-HABKJSAYSA-N | |
| 46185760 | |
| COC1=CC=C(C=C1)C2OCC3C(O2)C(C=CO3)O |
| C14H16O5 | |
| MFCD01863528 | |
| 4,6-o-4-methoxybenzylidene-d-glucal, 4 6-o-4-methoxybenzylidene-d-glucal, 4-o,6-o-4-methoxybenzylidene-1,2-dideoxy-d-arabino-1-hexenopyranose, 1,5-anhydro-4-o,6-o-4-methoxybenzylidene-2-deoxy-d-arabino-hexa-1-enitol, 4ar,8r,8as-2-4-methoxyphenyl-2h,4h,4ah,8h,8ah-pyrano 3,2-d 1,3 dioxin-8-ol | |
| (4aR,8R,8aS)-2-(4-methoxyphenyl)-4,4a,8,8a-tetrahydropyrano[3,2-d][1,3]dioxin-8-ol |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold