missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Nonadecadiyn-1-ol 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 mg |
| Formula Weight | 276.46 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4,6-Nonadecadiyn-1-ol |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |