accessibility menu, dialog, popup

UserName

H3COH

CAS RN 1071447-01-2

IUPAC Name: nonadeca-4,6-diyn-1-ol
Synonyms: nonadeca-4,6-diyn-1-ol
Molecular Weight (g/mol): 276.46
Molecular Formula: C19H32O
InChi Key: NQKQNTAHSMDCKX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC#CC#CCCCO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results