missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Hydroxyhexyloxy)-4'-methoxybiphenyl 95.0+%, TCI America™

Catalog No. H07671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H07671G Supplier TCI America Supplier No. H07671G

Chemical Identifiers

CAS 106869-53-8
Molecular Formula C19H24O3
Molecular Weight (g/mol) 300.398
MDL Number MFCD00191452
InChI Key DZAPEPIBAAMUIJ-UHFFFAOYSA-N
PubChem CID 44630340
IUPAC Name 6-[4-(4-methoxyphenyl)phenoxy]hexan-1-ol
SMILES COC1=CC=C(C=C1)C2=CC=C(C=C2)OCCCCCCO

Specifications

Melting Point 148°C
Color White
Quantity 1 g
Formula Weight 300.40
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(6-Hydroxyhexyloxy)-4′-methoxybiphenyl