accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 106869-53-8

H3COOOH

CAS RN 106869-53-8

IUPAC Name: 6-({4'-methoxy-[1,1'-biphenyl]-4-yl}oxy)hexan-1-ol
Synonyms: 6-({4'-methoxy-[1,1'-biphenyl]-4-yl}oxy)hexan-1-ol
Molecular Weight (g/mol): 300.4
Molecular Formula: C19H24O3
InChi Key: DZAPEPIBAAMUIJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(OCCCCCCO)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.