missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethylisophthalic Acid 95.0+%, TCI America™

Catalog No. D4377200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4377200MG 200 mg
D43771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4377200MG Supplier TCI America Supplier No. D4377200MG

Chemical Identifiers

CAS 2790-09-2
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
InChI Key HAYIPGIFANTODX-UHFFFAOYSA-N
PubChem CID 314208
IUPAC Name 4,6-dimethylbenzene-1,3-dicarboxylic acid
SMILES CC1=CC(=C(C=C1C(=O)O)C(=O)O)C

Specifications

Melting Point 298°C
Color White-Yellow
Quantity 200 mg
Formula Weight 194.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dimethylisophthalic Acid