accessibility menu, dialog, popup

UserName

CH3CH3OHOOHO

CAS RN 2790-09-2

IUPAC Name: 4,6-dimethylbenzene-1,3-dicarboxylic acid
Synonyms: 4,6-dimethylbenzene-1,3-dicarboxylic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: HAYIPGIFANTODX-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C1C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results