missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyldibenzothiophene, 95%

Catalog No. AC302190010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC302190010 1 g Glass bottle
AC302190050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC302190010 Supplier Thermo Scientific Chemicals Supplier No. 302190010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1207-12-1 | C14H12S | 212.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1207-12-1
Molecular Formula C14H12S
Molecular Weight (g/mol) 212.31
MDL Number MFCD00216264
InChI Key KMPJENUWHPZRGZ-UHFFFAOYSA-N
Synonym dibenzothiophene, 4,6-dimethyl, 4,6-dimethyldibenzo b,d thiophene, 4,6-dmdbt, 6,10-dimethyl-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4,6-dimethyl-dibenzothiophen, ksc491i3n, bidd:gt0307, 4,6-dimethyl dibenzo b thiophene, 4,6-dimethyldibenzothiophene, 4,6-dimethylbenzo b benzo b thiophene
PubChem CID 1268103
IUPAC Name 4,6-dimethyldibenzothiophene
SMILES CC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 147°C to 153°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ch2cl2
Formula Weight 212.31
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4, 6-Dimethyldibenzothiophene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-894-7

RUO – Research Use Only