accessibility menu, dialog, popup

UserName

H3CSH3C

CAS RN 1207-12-1

IUPAC Name: 3,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 3,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 212.31
Molecular Formula: C14H12S
InChi Key: KMPJENUWHPZRGZ-UHFFFAOYSA-N
SMILES: CC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)