missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dimethyl-2-(methylsulfonyl)pyrimidine 98.0+%, TCI America™
Supplier: TCI America D369925G
Specifications
| 4,6-Dimethyl-2-(methylsulfonyl)pyrimidine | |
| 82°C | |
| MFCD03788286 | |
| 4,6-dimethyl-2-methylsulfonyl pyrimidine, 2-methanesulfonyl-4,6-dimethylpyrimidine, 2-methanesulfonyl-4,6-dimethyl-pyrimidine, 2-methylsulfonyl-4,6-dimethylpyrimidine, pyrimidine, 4,6-dimethyl-2-methylsulfonyl, 4,6-dimethyl-2-methylsulfonyl-pyrimidine, pubchem10458, pubchem21462, acmc-209idk, ksc496i3d | |
| CC1=CC(=NC(=N1)S(=O)(=O)C)C | |
| 186.229 | |
| 186.23 | |
| Crystalline Powder |
| 35144-22-0 | |
| C7H10N2O2S | |
| 25 g | |
| ZHPSNGCLCHWTRG-UHFFFAOYSA-N | |
| 4,6-dimethyl-2-methylsulfonylpyrimidine | |
| 2772382 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 35144-22-0 | |
| 186.229 | |
| ZHPSNGCLCHWTRG-UHFFFAOYSA-N | |
| 2772382 | |
| CC1=CC(=NC(=N1)S(=O)(=O)C)C |
| C7H10N2O2S | |
| MFCD03788286 | |
| 4,6-dimethyl-2-methylsulfonyl pyrimidine, 2-methanesulfonyl-4,6-dimethylpyrimidine, 2-methanesulfonyl-4,6-dimethyl-pyrimidine, 2-methylsulfonyl-4,6-dimethylpyrimidine, pyrimidine, 4,6-dimethyl-2-methylsulfonyl, 4,6-dimethyl-2-methylsulfonyl-pyrimidine, pubchem10458, pubchem21462, acmc-209idk, ksc496i3d | |
| 4,6-dimethyl-2-methylsulfonylpyrimidine |
Safety and Handling
TSCA : No