missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyl-2-(methylsulfonyl)pyrimidine 98.0+%, TCI America™

Catalog No. p-7127872
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36995G 5 g
D369925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36995G Supplier TCI America Supplier No. D36995G

Chemical Identifiers

CAS 35144-22-0
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.229
MDL Number MFCD03788286
InChI Key ZHPSNGCLCHWTRG-UHFFFAOYSA-N
Synonym 4,6-dimethyl-2-methylsulfonyl pyrimidine, 2-methanesulfonyl-4,6-dimethylpyrimidine, 2-methanesulfonyl-4,6-dimethyl-pyrimidine, 2-methylsulfonyl-4,6-dimethylpyrimidine, pyrimidine, 4,6-dimethyl-2-methylsulfonyl, 4,6-dimethyl-2-methylsulfonyl-pyrimidine, pubchem10458, pubchem21462, acmc-209idk, ksc496i3d
PubChem CID 2772382
IUPAC Name 4,6-dimethyl-2-methylsulfonylpyrimidine
SMILES CC1=CC(=NC(=N1)S(=O)(=O)C)C

Specifications

Melting Point 82°C
Quantity 5 g
Formula Weight 186.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dimethyl-2-(methylsulfonyl)pyrimidine