missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyl-2-hydroxypyrimidine 98.0+%, TCI America™

Catalog No. d2427500g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D2427500G 500 g
D242725G 25 g
D2427100G 100 g
3 options

Catalog No. D2427500G

Supplier: TCI America D2427500G

Only null left

Chemical Identifiers

CAS 108-79-2
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD00006072
InChI Key WHEQVHAIRSPYDK-UHFFFAOYSA-N
Synonym 2-hydroxy-4,6-dimethylpyrimidine, 4,6-dimethylpyrimidin-2-ol, 4,6-dimethyl-2-hydroxypyrimidine, 4,6-dimethylpyrimidin-2 1h-one, 2 1h-pyrimidinone, 4,6-dimethyl, unii-hg0cbn4v9f, 4,6-dimethyl-2-pyrimidinol, 4,6-dimethyl-pyrimidin-2-ol, hg0cbn4v9f, 4,6-dimethyl-2-pyrimidone
PubChem CID 9508
IUPAC Name 4,6-dimethyl-1,2-dihydropyrimidin-2-one
SMILES CC1=CC(C)=NC(=O)N1

Specifications

Melting Point 202°C
Quantity 500 g
Formula Weight 124.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dimethyl-2-hydroxypyrimidine
EINECSNumber (9)-2245
TSCA Yes