missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dimethyl-2-hydroxypyrimidine 98.0+%, TCI America™
Supplier: TCI America D2427100G
Specifications
| 4,6-Dimethyl-2-hydroxypyrimidine | |
| 202°C | |
| MFCD00006072 | |
| 2-hydroxy-4,6-dimethylpyrimidine, 4,6-dimethylpyrimidin-2-ol, 4,6-dimethyl-2-hydroxypyrimidine, 4,6-dimethylpyrimidin-2 1h-one, 2 1h-pyrimidinone, 4,6-dimethyl, unii-hg0cbn4v9f, 4,6-dimethyl-2-pyrimidinol, 4,6-dimethyl-pyrimidin-2-ol, hg0cbn4v9f, 4,6-dimethyl-2-pyrimidone | |
| CC1=CC(C)=NC(=O)N1 | |
| 124.14 | |
| 124.14 | |
| Crystalline Powder |
| 108-79-2 | |
| C6H8N2O | |
| 100 g | |
| WHEQVHAIRSPYDK-UHFFFAOYSA-N | |
| 4,6-dimethyl-1,2-dihydropyrimidin-2-one | |
| 9508 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 108-79-2 | |
| 124.14 | |
| WHEQVHAIRSPYDK-UHFFFAOYSA-N | |
| 9508 | |
| CC1=CC(C)=NC(=O)N1 |
| C6H8N2O | |
| MFCD00006072 | |
| 2-hydroxy-4,6-dimethylpyrimidine, 4,6-dimethylpyrimidin-2-ol, 4,6-dimethyl-2-hydroxypyrimidine, 4,6-dimethylpyrimidin-2 1h-one, 2 1h-pyrimidinone, 4,6-dimethyl, unii-hg0cbn4v9f, 4,6-dimethyl-2-pyrimidinol, 4,6-dimethyl-pyrimidin-2-ol, hg0cbn4v9f, 4,6-dimethyl-2-pyrimidone | |
| 4,6-dimethyl-1,2-dihydropyrimidin-2-one |
Safety and Handling
EINECSNumber : (9)-2245
TSCA : Yes