missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dimethyl-2-hydroxypyrimidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America D1823250G
Specifications
| 4,6-Dimethyl-2-hydroxypyrimidine Hydrochloride | |
| 281°C | |
| MFCD00035312 | |
| 4,6-Dimethyl-2-pyrimidinol Hydrochloride, 2-Hydroxy-4,6-dimethylpyrimidine Hydrochloride | |
| CC1=CC(=NC(=O)N1)C.Cl | |
| 160.601 | |
| 160.60 | |
| Crystalline Powder |
| 34289-60-6 | |
| C6H9ClN2O | |
| 250 g | |
| IFXXETYYSLJRNF-UHFFFAOYSA-N | |
| 4,6-dimethyl-1H-pyrimidin-2-one;hydrochloride | |
| 12343166 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 34289-60-6 | |
| 160.601 | |
| IFXXETYYSLJRNF-UHFFFAOYSA-N | |
| 12343166 | |
| CC1=CC(=NC(=O)N1)C.Cl |
| C6H9ClN2O | |
| MFCD00035312 | |
| 4,6-Dimethyl-2-pyrimidinol Hydrochloride, 2-Hydroxy-4,6-dimethylpyrimidine Hydrochloride | |
| 4,6-dimethyl-1H-pyrimidin-2-one;hydrochloride |
Safety and Handling
EINECSNumber : (9)-2245
TSCA : No